Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289111.1 | NP_001276040.1 | 5 | 402 | ||
NM_032194.2 | NP_115570.1 | 6 | 440 | |||
XM_017011354.1 | XP_016866843.1 | 3 | 876 | |||
GenBank mRNA | AK024740.1 | - | 6 | 417 | ||
AK292165.1 | - | 6 | 419 | |||
AK301662.1 | - | 6 | 409 | |||
AL833613.1 | - | 5 | 370 | |||
BC035314.1 | - | 6 | 369 | 4745524 |