Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF562 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005259941.3 | XP_005259998.1 | 1 | 286 | ||
XM_017026898.1 | XP_016882387.1 | 1 | 286 |