Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARHGAP32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_005271736.4 | XP_005271793.2 | 4 | 1332 | ||
XM_011543072.2 | XP_011541374.1 | 4 | 534 | |||
XM_011543073.2 | XP_011541375.2 | 4 | 270 | |||
XM_017018595.1 | XP_016874084.1 | 4 | 251 | |||
XM_017018596.1 | XP_016874085.1 | 2 | 528 | |||
XM_017018597.1 | XP_016874086.1 | 4 | 1341 |