Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_011522085.2 | XP_011520387.1 | 1 | 709 | ||
XM_017022623.1 | XP_016878112.1 | 1 | 710 | |||
XM_017022624.1 | XP_016878113.1 | 1 | 709 | |||
XM_017022625.1 | XP_016878114.1 | 1 | 709 | |||
XM_017022626.1 | XP_016878115.1 | 1 | 709 | |||
XM_017022627.1 | XP_016878116.1 | 1 | 709 | |||
XM_017022628.1 | XP_016878117.1 | 1 | 709 | |||
XM_017022630.1 | XP_016878119.1 | 1 | 711 | |||
XM_017022631.1 | XP_016878120.1 | 1 | 710 | |||
XM_017022633.1 | XP_016878122.1 | 1 | 719 |