Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OR4F16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017002408.1 | XP_016857897.1 | 3 | 368 | ||
XM_017002409.1 | XP_016857898.1 | 4 | 286 | |||
XM_017002410.1 | XP_016857899.1 | 2 | 150 | |||
XM_017002411.1 | XP_016857900.1 | 2 | 139 |
Gene Symbol: LOC101927506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014132.1 | XP_016869621.1 | 4 | 3989 |
Gene Symbol: OR4F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017009338.1 | XP_016864827.1 | 2 | 1851 |