Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC146880 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027487.2 | 3 | 1543 | |||
GenBank mRNA | AK097700.1 | - | 1 | 873 | ||
BC032382.1 | - | 3 | 1543 |