Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00266-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040415.1 | 2, 3 | 520 | |||
GenBank mRNA | BC118988.1 | - | 2, 3 | 520 | 40035995 | |
BC122537.1 | - | 2, 3 | 520 | 40035996 |
Gene Symbol: LOC101927506 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014131.1 | XP_016869620.1 | 2, 3 | 2097 | ||
XM_017014132.1 | XP_016869621.1 | 2, 3 | 2097 | |||
XM_017014133.1 | XP_016869622.1 | 2, 3 | 2097 |