Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTOR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004958.3 | NP_004949.1 | 46 | 6579 | ||
XM_005263438.2 | XP_005263495.1 | 46 | 6586 | |||
XM_017000900.1 | XP_016856389.1 | 43 | 6042 | |||
XM_017000901.1 | XP_016856390.1 | 44 | 6330 | |||
GenBank mRNA | AB209995.1 | - | 46 | 6560 | ||
AK126762.1 | - | 8 | 1875 | |||
BC117166.1 | - | 45 | 6476 | 40125717 | ||
L34075.1 | - | 46 | 6537 | |||
U88966.1 | - | 46 | 6534 |