Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NNT-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_073113.1 | 1 | 215 | |||
GenBank mRNA | DA630653.1 | - | 1 | 215 | ||
DB499358.1 | - | 1 | 121 | |||
HY014991.1 | - | 1 | 130 |