Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024630.2 | NP_078906.2 | 3 | 1051 | ||
NM_153746.1 | NP_714968.1 | 3 | 1051 | |||
XM_005267146.3 | XP_005267203.1 | 2 | 422 | |||
XM_011536120.2 | XP_011534422.1 | 3 | 1632 | |||
XM_011536123.2 | XP_011534425.1 | 3 | 443 | |||
XM_017011307.1 | XP_016866796.1 | 3 | 736 | |||
XM_017011308.1 | XP_016866797.1 | 3 | 736 | |||
XM_017011309.1 | XP_016866798.1 | 3 | 736 | |||
XM_017011310.1 | XP_016866799.1 | 3 | 736 | |||
XM_017011311.1 | XP_016866800.1 | 3 | 736 | |||
XM_017011312.1 | XP_016866801.1 | 3 | 751 | |||
XM_017011313.1 | XP_016866802.1 | 4 | 3448 | |||
GenBank mRNA | AF542388.1 | - | 3 | 1051 | ||
AF542389.1 | - | 3 | 1051 | |||
AK309408.1 | - | 2 | 305 | |||
BC029881.1 | - | 2 | 165 | |||
BC125072.1 | - | 4 | 568 | |||
BC125073.1 | - | 4 | 568 |