Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZDHHC14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024630.2 | NP_078906.2 | 7 | 1411 | ||
NM_153746.1 | NP_714968.1 | 7 | 1411 | |||
XM_005267146.3 | XP_005267203.1 | 6 | 782 | |||
XM_011536120.2 | XP_011534422.1 | 7 | 1992 | |||
XM_011536123.2 | XP_011534425.1 | 7 | 803 | |||
XM_017011307.1 | XP_016866796.1 | 7 | 1096 | |||
XM_017011308.1 | XP_016866797.1 | 7 | 1096 | |||
XM_017011309.1 | XP_016866798.1 | 7 | 1096 | |||
XM_017011310.1 | XP_016866799.1 | 7 | 1096 | |||
XM_017011311.1 | XP_016866800.1 | 6 | 958 | |||
XM_017011312.1 | XP_016866801.1 | 7 | 1111 | |||
XM_017011313.1 | XP_016866802.1 | 8 | 3808 | |||
GenBank mRNA | AF542388.1 | - | 7 | 1411 | ||
AF542389.1 | - | 7 | 1411 | |||
AK024637.1 | - | 3 | 220 | |||
AK092380.1 | - | 3 | 1922 | |||
AK124573.1 | - | 4 | 3778 | |||
AK128773.1 | - | 3 | 2730 | |||
BC008978.2 | - | 1 | 182 | |||
BC029881.1 | - | 6 | 525 | |||
BC056399.1 | - | 1 | 53 | |||
BC125072.1 | - | 8 | 928 | |||
BC125073.1 | - | 8 | 928 |