Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPR35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195381.1 | NP_001182310.1 | 6 | 1140 | ||
NM_001195382.1 | NP_001182311.1 | 6 | 1026 | |||
NM_005301.3 | NP_005292.2 | 2 | 503 | |||
GenBank mRNA | AF089087.1 | - | 1 | 750 | ||
AK131540.1 | - | 1 | 2686 | |||
AK172786.1 | - | 6 | 1117 | |||
AK290256.1 | - | 6 | 989 | |||
AK314002.1 | - | 2 | 503 | |||
BC095500.1 | - | 1 | 172 | |||
BC117453.1 | - | 1 | 547 | 40126004 | ||
BC117455.1 | - | 1 | 545 | 40126006 | ||
CR541765.1 | - | 1 | 175 |