Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM229A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136002.1 | NP_001129474.1 | 1 | 635 | ||
GenBank mRNA | BC157828.1 | - | 1 | 189 | 9057047 | |
BC171809.1 | - | 1 | 189 | 9054463 |