Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FFAR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005304.3 | NP_005295.1 | 2 | 1061 | ||
XM_011526858.2 | XP_011525160.1 | 2 | 1417 | |||
GenBank mRNA | AK290710.1 | - | 2 | 1061 | ||
BC035657.1 | - | 2 | 1047 | 5588752 | ||
BC113695.1 | - | 1 | 1056 | 8322747 | ||
BC148269.1 | - | 1 | 1008 | 8860212 | ||
EU432115.1 | - | 1 | 986 |