Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKIDA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207371.3 | NP_997254.3 | 4 | 2445 | ||
XM_005252450.3 | XP_005252507.1 | 5 | 1554 | |||
XM_011519480.2 | XP_011517782.1 | 5 | 1551 | |||
XM_017016225.1 | XP_016871714.1 | 5 | 2345 | |||
XM_017016226.1 | XP_016871715.1 | 5 | 2348 | |||
GenBank mRNA | AB234866.1 | - | 1 | 289 | ||
AK131456.1 | - | 4 | 2445 | |||
AK303207.1 | - | 2 | 462 |