Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKIDA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207371.3 | NP_997254.3 | 4 | 3787 | ||
XM_005252450.3 | XP_005252507.1 | 5 | 2896 | |||
XM_011519480.2 | XP_011517782.1 | 5 | 2893 | |||
XM_017016225.1 | XP_016871714.1 | 5 | 3687 | |||
XM_017016226.1 | XP_016871715.1 | 5 | 3690 | |||
GenBank mRNA | AB234866.1 | - | 2 | 1388 | ||
AK127130.1 | - | 2 | 822 | |||
AK131456.1 | - | 5 | 3523 | |||
AK303207.1 | - | 3 | 1484 | |||
AL834446.1 | - | 1 | 505 |