Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282873.1 | NP_001269802.1 | 30 | 4353 | ||
NM_015902.5 | NP_056986.2 | 30 | 4353 | |||
XM_005250962.4 | XP_005251019.1 | 9 | 1114 | |||
XM_011517104.2 | XP_011515406.2 | 29 | 4338 | |||
XM_011517105.2 | XP_011515407.2 | 29 | 4338 | |||
XM_011517106.2 | XP_011515408.2 | 29 | 4320 | |||
XM_017013533.1 | XP_016869022.1 | 29 | 4338 | |||
XM_017013534.1 | XP_016869023.1 | 29 | 4338 | |||
GenBank mRNA | AB020703.3 | - | 30 | 4353 | ||
AF006010.2 | - | 30 | 3931 | |||
BC137234.1 | - | 30 | 3925 | 9021231 | ||
U95000.1 | - | 30 | 3895 |