Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRK6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004105.2 | NP_001004105.1 | 6 | 659 | ||
NM_001004106.2 | NP_001004106.1 | 6 | 659 | |||
NM_002082.3 | NP_002073.2 | 6 | 659 | |||
XM_006714858.3 | XP_006714921.1 | 4 | 1300 | |||
XM_006714859.2 | XP_006714922.1 | 6 | 571 | |||
XM_011534537.2 | XP_011532839.1 | 5 | 411 | |||
GenBank mRNA | AF040751.1 | - | 6 | 502 | ||
AF040752.1 | - | 6 | 502 | |||
AK056697.1 | - | 4 | 637 | |||
BC009277.2 | - | 6 | 599 | 4053197 | ||
BC017272.1 | - | 6 | 548 | 4640283 | ||
BU507609.1 | - | 6 | 658 | |||
BX382886.2 | - | 6 | 657 | |||
L16862.1 | - | 6 | 561 |
Gene Symbol: GRK6P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK295462.1 | - | 1 | 564 |