Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H3F3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002107.4 | NP_002098.1 | 4 | 1039 | ||
GenBank mRNA | BC013857.1 | - | 1 | 907 | ||
BC038989.1 | - | 1 | 939 | 6014954 | ||
BC081561.1 | - | 1 | 1011 | 5162817 | ||
M11353.1 | - | 3 | 1283 | |||
M11354.1 | - | 4 | 1017 |
Gene Symbol: H3F3AP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_002315.1 | 1 | 1018 | |||
GenBank mRNA | AK291102.1 | - | 4 | 1020 | ||
BC066901.1 | - | 1 | 1014 | |||
BC067757.1 | - | 1 | 1020 | |||
BC081560.1 | - | 1 | 1011 | 3884607 |