Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001017405.2 | NP_001017405.1 | 9 | 1961 | ||
NM_001297430.1 | NP_001284359.1 | 8 | 1757 | |||
NM_001297431.1 | NP_001284360.1 | 7 | 1757 | |||
NM_001297432.1 | NP_001284361.1 | 9 | 2099 | |||
NM_001297433.1 | NP_001284362.1 | 9 | 2035 | |||
NM_005882.4 | NP_005873.2 | 8 | 1838 | |||
NR_123716.1 | 7 | 1761 | ||||
XM_017007636.1 | XP_016863125.1 | 10 | 2081 | |||
XM_017007637.1 | XP_016863126.1 | 9 | 1998 | |||
GenBank mRNA | AB208907.1 | - | 3 | 5117 | ||
AF084928.1 | - | 10 | 1755 | |||
AK001088.1 | - | 9 | 1921 | |||
AK022515.1 | - | 7 | 1726 | |||
AK022586.1 | - | 8 | 1798 | |||
AK091951.1 | - | 4 | 1832 | |||
AK094253.1 | - | 6 | 1582 | |||
AK125501.1 | - | 1 | 2213 | |||
AK127875.1 | - | 5 | 2189 | |||
AK128302.1 | - | 3 | 2865 | |||
BC001225.1 | - | 9 | 1928 | 3533990 | ||
BC006470.1 | - | 9 | 1884 |