Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TATDN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146160.1 | NP_001139632.1 | 3 | 191 | ||
NM_001317889.1 | NP_001304818.1 | 5 | 307 | |||
NM_001317890.1 | NP_001304819.1 | 3 | 290 | |||
NM_032026.3 | NP_114415.1 | 5 | 307 | |||
NR_027427.1 | 5 | 307 | ||||
XM_006716666.3 | XP_006716729.1 | 5 | 327 | |||
XM_006716667.3 | XP_006716730.1 | 5 | 372 | |||
XM_006716668.3 | XP_006716731.1 | 5 | 385 | |||
XM_006716669.3 | XP_006716732.1 | 3 | 269 | |||
XM_006716670.3 | XP_006716733.1 | 4 | 228 | |||
XM_006716671.3 | XP_006716734.1 | 4 | 306 | |||
XM_011517331.2 | XP_011515633.1 | 4 | 223 | |||
XM_017013895.1 | XP_016869384.1 | 5 | 317 | |||
XM_017013896.1 | XP_016869385.1 | 5 | 372 | |||
XM_017013897.1 | XP_016869386.1 | 4 | 293 | |||
XM_017013898.1 | XP_016869387.1 | 4 | 230 | |||
XM_017013899.1 | XP_016869388.1 | 5 | 303 | |||
GenBank mRNA | AF212250.1 | - | 5 | 290 | ||
AK312204.1 | - | 5 | 303 | |||
AY071865.1 | - | 3 | 180 | |||
BC064964.1 | - | 5 | 291 | 6141834 | ||
BG532054.1 | - | 3 | 280 | |||
BX332564.2 | - | 5 | 289 |