Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002083.3 | NP_002074.2 | 2 | 699 | ||
NR_046320.1 | 3 | 905 | ||||
NR_046321.1 | 3 | 904 | ||||
GenBank mRNA | AK225871.1 | - | 3 | 746 | ||
BC005277.1 | - | 2 | 539 | 3996732 | ||
BC016756.1 | - | 2 | 553 | 3681457 | ||
BC022820.1 | - | 2 | 522 | 5429926 | ||
BC067221.1 | - | 2 | 543 | 6668339 | ||
X53463.1 | - | 2 | 571 | |||
X68314.1 | - | 2 | 554 |