Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039847.2 | NP_001034936.1 | 3 | 347 | ||
NM_001039848.2 | NP_001034937.1 | 3 | 425 | |||
NM_002085.4 | NP_002076.2 | 3 | 347 | |||
GenBank mRNA | AK225859.1 | - | 3 | 266 | ||
BC010157.2 | - | 3 | 175 | |||
BC011836.2 | - | 3 | 268 | 3945177 | ||
BC021567.1 | - | 3 | 261 | 4581069 | ||
BC022071.1 | - | 3 | 284 | 4821595 | ||
BC032695.1 | - | 3 | 268 | 5227887 | ||
BC039849.1 | - | 3 | 336 | 5744771 | ||
BC046163.1 | - | 3 | 243 | 5742889 | ||
BC058329.1 | - | 3 | 264 | |||
BI561548.1 | - | 3 | 343 | |||
BM764133.1 | - | 3 | 426 | |||
BQ648717.1 | - | 3 | 212 | |||
CF127510.1 | - | 3 | 359 | |||
HY049693.1 | - | 3 | 348 | |||
X71973.1 | - | 3 | 309 |