Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030002.2 | NP_001025173.1 | 8, 9 | 1039 | ||
NM_001242442.1 | NP_001229371.1 | 8, 9 | 1123 | |||
NM_001242443.1 | NP_001229372.1 | 8, 9 | 1184 | |||
NM_005310.3 | NP_005301.2 | 8, 9 | 1194 | |||
XM_017024536.1 | XP_016880025.1 | 7, 8 | 2313 | |||
XM_017024537.1 | XP_016880026.1 | 8, 9 | 1161 | |||
XM_017024538.1 | XP_016880027.1 | 7, 8 | 1361 | |||
GenBank mRNA | AB008789.1 | - | 7, 8 | 970 | ||
AB008790.1 | - | 7, 8 | 970 | |||
AK027729.1 | - | 8, 9 | 1039 | |||
AK125544.1 | - | 8, 9 | 1123 | |||
AK222849.1 | - | 8, 9 | 1142 | |||
AK222870.1 | - | 8, 9 | 1141 | |||
AK290115.1 | - | 8, 9 | 1184 | |||
AK314368.1 | - | 8, 9 | 1176 | |||
BC006535.2 | - | 8, 9 | 1025 | 2824302 | ||
BT006686.1 | - | 7, 8 | 919 | |||
D43772.1 | - | 8, 9 | 1138 | |||
D87513.1 | - | 7, 8 | 850 | |||
KU178103.1 | - | 7, 8 | 919 | |||
KU178104.1 | - | 6, 7 | 1044 |