Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERBB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005862.2 | NP_001005862.1 | 26, 27 | 3367 | ||
NM_001289936.1 | NP_001276865.1 | 27, 28 | 3418 | |||
NM_001289937.1 | NP_001276866.1 | 23, 24 | 3142 | |||
NM_004448.3 | NP_004439.2 | 23, 24 | 3142 | |||
NR_110535.1 | 23, 24 | 3205 | ||||
GenBank mRNA | AK295195.1 | - | 27, 28 | 3418 | ||
AK300321.1 | - | 17, 18 | 2316 | |||
AK313683.1 | - | 23, 24 | 3096 | |||
BC080193.1 | - | 23, 24 | 3103 | |||
BC167147.1 | - | 23, 24 | 2884 | 100013444 | ||
KJ534831.1 | - | 23, 24 | 2881 | |||
M11730.1 | - | 23, 24 | 3031 | |||
X03363.1 | - | 23, 24 | 3055 |