Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOP3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282112.1 | NP_001269041.1 | 18 | 2469 | ||
NM_001282113.1 | NP_001269042.1 | 18 | 2462 | |||
NM_003935.4 | NP_003926.1 | 18 | 2785 | |||
XM_005261811.1 | XP_005261868.1 | 20 | 2635 | |||
XM_005261813.2 | XP_005261870.1 | 19 | 2500 | |||
XM_006724349.1 | XP_006724412.1 | 19 | 2446 | |||
XM_006724350.1 | XP_006724413.1 | 19 | 2620 | |||
XM_011530482.1 | XP_011528784.1 | 19 | 2702 | |||
XM_017029039.1 | XP_016884528.1 | 20 | 2510 | |||
XM_017029040.1 | XP_016884529.1 | 18 | 3382 | |||
XM_017029041.1 | XP_016884530.1 | 19 | 3424 | |||
GenBank mRNA | AF017146.1 | - | 17 | 2370 | ||
AF053082.1 | - | 17 | 2370 | |||
AF125216.1 | - | 17 | 2330 | |||
AK096695.1 | - | 10 | 2475 | |||
AK290997.1 | - | 18 | 2415 | |||
AL833505.1 | - | 17 | 3749 | |||
BC002432.2 | - | 18 | 2692 | 3346377 | ||
CR456596.1 | - | 17 | 2284 |