Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COL6A4P1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027927.1 | 2 | 475 | |||
GenBank mRNA | AB299979.1 | - | 2 | 475 | ||
FN394065.1 | - | 2 | 309 |