Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000829.3 | NP_000820.3 | 4 | 749 | ||
NM_001077243.2 | NP_001070711.2 | 4 | 749 | |||
NM_001077244.1 | NP_001070712.1 | 4 | 749 | |||
NM_001112812.1 | NP_001106283.1 | 3 | 617 | |||
NR_046356.1 | 3 | 617 | ||||
XM_005271518.3 | XP_005271575.1 | 4 | 793 | |||
XM_006718823.1 | XP_006718886.1 | 4 | 423 | |||
XM_011542775.2 | XP_011541077.1 | 4 | 793 | |||
XM_017017609.1 | XP_016873098.1 | 4 | 793 | |||
XM_017017610.1 | XP_016873099.1 | 4 | 793 | |||
GenBank mRNA | AB209092.1 | - | 3 | 587 | ||
BC045546.1 | - | 4 | 743 | |||
BC114549.1 | - | 3 | 302 | |||
BC142654.1 | - | 4 | 697 | |||
BC146816.1 | - | 3 | 576 | |||
BC150209.1 | - | 4 | 697 | |||
U16129.1 | - | 3 | 303 |