Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK000031.1 | - | 9 | 966 | ||
AK027029.1 | - | 7 | 1119 | |||
AK293136.1 | - | 10 | 1169 | |||
AK293365.1 | - | 8 | 883 | |||
AK297633.1 | - | 8 | 837 | |||
AK297971.1 | - | 9 | 918 | |||
BC022271.1 | - | 9 | 951 | 4688412 |
Gene Symbol: FAM45BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF220185.1 | - | 1 | 1158 |