Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRID2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286838.1 | NP_001273767.1 | 14 | 2535 | ||
NM_001510.3 | NP_001501.2 | 15 | 2820 | |||
XM_011531893.2 | XP_011530195.2 | 15 | 9057 | |||
XM_011531894.2 | XP_011530196.1 | 13 | 2096 | |||
XM_011531895.2 | XP_011530197.1 | 13 | 2013 | |||
XM_017008118.1 | XP_016863607.1 | 14 | 9009 | |||
XM_017008119.1 | XP_016863608.1 | 15 | 2594 | |||
XM_017008120.1 | XP_016863609.1 | 15 | 9057 | |||
XM_017008123.1 | XP_016863612.1 | 13 | 3324 | |||
XM_017008124.1 | XP_016863613.1 | 12 | 2544 | |||
XM_017008125.1 | XP_016863614.1 | 12 | 2041 | |||
XM_017008126.1 | XP_016863615.1 | 11 | 1757 | |||
GenBank mRNA | AB209318.1 | - | 15 | 2820 | ||
AF009014.1 | - | 15 | 2562 | |||
BC099652.3 | - | 15 | 2562 | 40008396 | ||
BC099653.3 | - | 15 | 2562 | 40008397 | ||
BC099654.3 | - | 14 | 2277 | 40008398 |