Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRBOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205272.1 | NP_001192201.1 | 5 | 478 | ||
GenBank mRNA | AK130974.1 | - | 5 | 460 | ||
BC062786.1 | - | 5 | 464 | |||
BC103985.1 | - | 4 | 523 | |||
BC103986.1 | - | 4 | 523 | |||
BC103987.1 | - | 4 | 523 | |||
BC118551.1 | - | 4 | 523 | |||
BC118662.1 | - | 4 | 523 | |||
DB458286.2 | - | 5 | 460 |