Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WHAMMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_036650.1 | 3 | 760 | |||
GenBank mRNA | BC117556.1 | - | 760 | |||
BI910970.1 | - | 2 | 196 |
Gene Symbol: WHAMMP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_026589.1 | 5 | 1161 | |||
GenBank mRNA | BC035099.1 | - | 5 | 711 | ||
BC066982.1 | - | 5 | 885 | |||
BC150524.1 | - | 3 | 390 |
Gene Symbol: WHAMMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003521.1 | 5 | 1137 | |||
GenBank mRNA | BC018097.1 | - | 4 | 456 | ||
BC044610.1 | - | 4 | 589 | |||
BC048987.1 | - | 5 | 1197 | |||
BC093705.1 | - | 2 | 194 | |||
DQ309039.1 | - | 4 | 446 | |||
DQ309040.1 | - | 5 | 1137 |