Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MUC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005960.1 | NP_005951.1 | 2 | 2181 | ||
XM_011516246.2 | XP_011514548.1 | 2 | 2540 | |||
XM_011516247.2 | XP_011514549.1 | 2 | 2541 | |||
XM_011516249.2 | XP_011514551.1 | 2 | 2545 | |||
XM_011516250.2 | XP_011514552.1 | 2 | 2540 | |||
XM_017012231.1 | XP_016867720.1 | 2 | 2547 |