Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VRK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288837.1 | NP_001275766.1 | 2 | 248 | ||
NM_001288838.1 | NP_001275767.1 | 2 | 248 | |||
GenBank mRNA | AK292727.1 | - | 2 | 234 | ||
AK307113.1 | - | 3 | 318 |