Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001272101.1 | NP_001259030.1 | 7 | 1252 | ||
NM_002429.5 | NP_002420.1 | 9 | 1520 | |||
NR_073606.1 | 9 | 1597 | ||||
XM_006719401.3 | XP_006719464.1 | 7 | 1161 | |||
XM_011538359.1 | XP_011536661.1 | 8 | 1260 | |||
XM_017019309.1 | XP_016874798.1 | 8 | 1378 | |||
GenBank mRNA | AK225939.1 | - | 6 | 834 | ||
AK303202.1 | - | 7 | 1205 | |||
BC050368.1 | - | 9 | 1459 | 5761074 | ||
U37791.1 | - | 9 | 1448 | |||
U38320.1 | - | 3 | 2048 | |||
U38321.1 | - | 9 | 1443 | |||
U38322.1 | - | 5 | 1254 | |||
U38431.1 | - | 9 | 1518 | |||
X92521.1 | - | 9 | 1468 | |||
Y08622.1 | - | 9 | 1467 |