Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUTM2D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_075100.1 | 2, 3 | 1415 | |||
GenBank mRNA | AB095940.1 | - | 2, 3 | 1415 |
Gene Symbol: NUTM2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099338.1 | NP_001092808.1 | 2, 3 | 1468 | ||
XM_011540122.2 | XP_011538424.1 | 4, 5 | 4311 | |||
XM_017016607.1 | XP_016872096.1 | 4, 5 | 4312 | |||
GenBank mRNA | BC092519.1 | - | 2, 3 | 1464 |
Gene Symbol: NUTM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278495.1 | NP_001265424.1 | 2, 3 | 1468 |
Gene Symbol: NUTM2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017561.1 | NP_060031.1 | 2, 3 | 737 | ||
GenBank mRNA | AL133071.1 | - | 1, 2 | 290 | ||
BC130390.1 | - | 2, 3 | 737 | 40146421 | ||
BC143798.1 | - | 2, 3 | 737 | 9052312 |
Gene Symbol: NUTM2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001045477.2 | NP_001038942.1 | 2, 3 | 737 | ||
NM_001170741.1 | NP_001164212.1 | 2, 3 | 737 |