Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aatk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198785.1 | NP_001185714.1 | 13 | 4246 | ||
NM_001198787.1 | NP_001185716.1 | 13 | 4018 | |||
NM_007377.4 | NP_031403.2 | 13 | 4529 | |||
GenBank mRNA | AB093253.2 | - | 13 | 4529 | ||
AB288871.1 | - | 13 | 4148 | |||
AF011908.1 | - | 13 | 3985 | |||
AK036705.1 | - | 13 | 4020 | |||
AK147239.1 | - | 10 | 4039 | |||
AK155100.1 | - | 13 | 4020 | |||
AY236858.1 | - | 13 | 4024 | |||
AY236859.1 | - | 13 | 4051 | |||
BC042837.1 | - | 13 | 4459 | |||
BC052323.1 | - | 13 | 4460 | |||
BC080846.1 | - | 13 | 4100 | 6856191 |