Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aatk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198785.1 | NP_001185714.1 | 11 | 3520 | ||
NM_001198787.1 | NP_001185716.1 | 11 | 3292 | |||
NM_007377.4 | NP_031403.2 | 11 | 3803 | |||
GenBank mRNA | AB093253.2 | - | 11 | 3803 | ||
AB288871.1 | - | 11 | 3422 | |||
AF011908.1 | - | 11 | 3259 | |||
AK036705.1 | - | 11 | 3294 | |||
AK147239.1 | - | 8 | 3313 | |||
AK155100.1 | - | 11 | 3294 | |||
AY236858.1 | - | 11 | 3298 | |||
AY236859.1 | - | 11 | 3325 | |||
BC042837.1 | - | 11 | 3733 | |||
BC052323.1 | - | 11 | 3734 | |||
BC080846.1 | - | 11 | 3374 | 6856191 |