Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112703.2 | NP_001106174.1 | 3 | 1632 | ||
NM_001283045.1 | NP_001269974.1 | 3 | 509 | |||
NM_001283046.1 | NP_001269975.1 | 4 | 1837 | |||
NM_001283047.1 | NP_001269976.1 | 4 | 1776 | |||
NM_009594.4 | NP_033724.2 | 3 | 403 | |||
GenBank mRNA | AK088252.1 | - | 4 | 1762 | ||
AK090095.1 | - | 3 | 361 | |||
AK166028.1 | - | 3 | 1621 | |||
AK166210.1 | - | 4 | 1871 | |||
BC059260.1 | - | 3 | 430 | 6408990 | ||
BC103770.1 | - | 3 | 856 | 30848553 | ||
CJ099623.1 | - | 3 | 354 | |||
CJ128107.1 | - | 3 | 398 | |||
CN663033.1 | - | 2 | 124 | |||
J02995.1 | - | 3 | 356 | |||
K03228.1 | - | 3 | 398 | |||
M12263.1 | - | 4 | 763 | |||
M12264.1 | - | 3 | 509 | |||
M12265.1 | - | 3 | 558 |