Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001112703.2 | NP_001106174.1 | 6 | 2256 | ||
NM_001283045.1 | NP_001269974.1 | 6 | 1133 | |||
NM_001283046.1 | NP_001269975.1 | 7 | 2461 | |||
NM_001283047.1 | NP_001269976.1 | 7 | 2400 | |||
NM_009594.4 | NP_033724.2 | 6 | 1027 | |||
GenBank mRNA | AK088252.1 | - | 7 | 2386 | ||
AK090095.1 | - | 6 | 985 | |||
AK166028.1 | - | 6 | 2245 | |||
AK218428.1 | - | 3 | 144 | |||
BC059260.1 | - | 6 | 1054 | 6408990 | ||
BC103770.1 | - | 6 | 1480 | 30848553 | ||
CF727094.1 | - | 3 | 160 | |||
CK626997.1 | - | 3 | 179 | |||
J02995.1 | - | 6 | 980 | |||
L10656.1 | - | 4 | 581 |