Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009601.4 | NP_033731.3 | 6 | 645 | ||
GenBank mRNA | AK087554.1 | - | 6 | 624 | ||
BC023318.1 | - | 6 | 651 | 4009493 | ||
M14537.1 | - | 6 | 588 |