Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009602.4 | NP_033732.2 | 5 | 1526 | ||
GenBank mRNA | AF145286.1 | - | 5 | 1465 | ||
AF299083.1 | - | 5 | 1320 | |||
AK049722.1 | - | 5 | 1499 | |||
AK051742.1 | - | 5 | 1508 | |||
AK081254.1 | - | 1503 | ||||
AY574267.1 | - | 5 | 1300 | |||
BC065103.1 | - | 5 | 1479 | 6838257 |