Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009602.4 | NP_033732.2 | 4 | 495 | ||
GenBank mRNA | AF089739.1 | - | 3 | 185 | ||
AF145286.1 | - | 4 | 434 | |||
AF299083.1 | - | 4 | 289 | |||
AF325349.1 | - | 4 | 255 | |||
AK049722.1 | - | 4 | 468 | |||
AK051742.1 | - | 4 | 477 | |||
AK081254.1 | - | 472 | ||||
AY574267.1 | - | 4 | 269 | |||
BC065103.1 | - | 4 | 448 | 6838257 |