Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009604.3 | NP_033734.3 | 10 | 1213 | ||
GenBank mRNA | AK047974.1 | - | 10 | 1214 | ||
M30514.1 | - | 10 | 1254 | |||
X03818.1 | - | 10 | 1210 |