Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278064.1 | NP_001264993.1 | 7 | 2457 | ||
NM_001278065.1 | NP_001264994.1 | 8 | 2504 | |||
NM_001278066.1 | NP_001264995.1 | 7 | 2457 | |||
NM_001278067.1 | NP_001264996.1 | 7 | 2457 | |||
XM_011535782.1 | XP_011534084.1 | 8 | 2607 | |||
XM_017010783.1 | XP_016866272.1 | 8 | 2616 | |||
XM_017010784.1 | XP_016866273.1 | 8 | 2652 | |||
XM_017010785.1 | XP_016866274.1 | 8 | 2612 | |||
XM_017010786.1 | XP_016866275.1 | 8 | 2612 | |||
XM_017010787.1 | XP_016866276.1 | 8 | 2648 | |||
XM_017010788.1 | XP_016866277.1 | 6 | 1424 | |||
GenBank mRNA | AB208837.1 | - | 7 | 2124 | ||
BC136280.1 | - | 7 | 2169 | 9020267 | ||
BC143777.1 | - | 7 | 2169 | |||
BC143779.1 | - | 7 | 2167 | |||
L76627.1 | - | 7 | 2137 | |||
L76631.1 | - | 8 | 2507 | |||
U31215.1 | - | 7 | 2372 | |||
U31216.1 | - | 7 | 2137 |
Set Membership: |
G-protein coupled receptor |