Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033268.4 | NP_150371.4 | 21 | 2815 | ||
GenBank mRNA | AF248643.1 | - | 21 | 2583 | ||
AK003570.1 | - | 5 | 578 | |||
AY036877.1 | - | 22 | 2759 | |||
BC089579.1 | - | 21 | 2815 | 6757718 |