Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Actn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033268.4 | NP_150371.4 | 10 | 1212 | ||
GenBank mRNA | AF248643.1 | - | 10 | 980 | ||
AY036877.1 | - | 11 | 1156 | |||
BC089579.1 | - | 10 | 1212 | 6757718 |