Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007399.3 | NP_031425.2 | 7 | 996 | ||
GenBank mRNA | AF011379.1 | - | 7 | 817 | ||
AK036599.1 | - | 1 | 40 | |||
BC057346.1 | - | 7 | 958 | |||
BC066207.1 | - | 7 | 1001 |