Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007402.2 | NP_031428.2 | 17 | 1870 | ||
GenBank mRNA | AF013107.1 | - | 17 | 1856 | ||
AK136692.1 | - | 17 | 1869 | |||
AK164965.1 | - | 3 | 390 |