Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adam8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089086.1 | - | 17 | 1841 | ||
AK089169.1 | - | 17 | 1881 | |||
AK134118.1 | - | 7 | 1395 | |||
AK150411.1 | - | 17 | 1843 | |||
AK151535.1 | - | 9 | 1072 | |||
AK152675.1 | - | 17 | 1860 | |||
AK152769.1 | - | 17 | 1842 | |||
AK153065.1 | - | 17 | 1842 | |||
AK155772.1 | - | 17 | 1841 | |||
AK155912.1 | - | 17 | 1861 | |||
AK160041.1 | - | 17 | 1844 | |||
AK170019.1 | - | 17 | 1866 | |||
AK171022.1 | - | 15 | 2418 | |||
AK171060.1 | - | 17 | 1841 | |||
BC025584.1 | - | 17 | 1895 | 5324943 | ||
X13335.1 | - | 17 | 1840 |